Learn More
Perkin Elmer US LLC GC/MS PSS Injector Starter Kit
Supplier: Perkin Elmer US LLC N6100447
Kit includes the following pieces: (1) 5.0 µL Autosampler Syringe (2) Vial Locator (dongle) (10) PSS Injector Viton O-Rings (250°C) (1) PSS Injector Kelrez O-Rings (350°C) (1) PSS Split/Splitless Injector, 2 mm, No Wool (20) Graphite/Vespel Ferrules, for 0.25 mm Columns (1) PerkinElmer Green Septa (50 pieces) (1) Marathon Filament (1) Aluminum Oxide Powder (3 oz.)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.