Learn More
Perkin Elmer US LLC GC Quiet Compressor - 220 V, 60 Hz
Supplier: Perkin Elmer US LLC N9306292
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This compressor is quiet and lightweight for use in the laboratory area. It contains a 5.0 micron pre-filter/regulator and an internally powder-coated air tank to prevent rust along with a pressure switch for automatic operation. It comes with a 1 year warranty on workmanship. Power Requirements: 220 V/60 Hz Horsepower: 0.60 Output: 2.5 CFM/67 L/min Operating Pressure: 80-100 psi/6-7 bar Maximum Pressure: 100 psi/7 bar Noise Level: 62 db/A Tank Size: 24 L/6.0 Gal Dimensions: 16 in x 16 in x 20 in (Packed: 19 in x 18 in x 23 in) Weight: 54 lbs (Packed: 58 lbs)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.