Learn More
Perkin Elmer US LLC GC Quiet Compressor - 220 V, 50 Hz
Supplier:  Perkin Elmer US LLC N9306293
Encompass_Preferred
This compressor is quiet and lightweight for use in the laboratory area. It contains a 5.0 micron pre-filter/regulator and an internally powder-coated air tank to prevent rust along with a pressure switch for automatic operation. It comes with a 1 year warranty on workmanship. Power Requirements: 220 V/50 Hz Horsepower: 0.60 Output: 2.5 CFM/67 L/min Operating Pressure: 80-100 psi/6-7 bar Maximum Pressure: 100 psi/7 bar Noise Level: 62 db/A Tank Size: 24 L/6.0 Gal Dimensions: 16 in x 16 in x 20 in (Packed: 19 in x 18 in x 23 in) Weight: 54 lbs (Packed: 58 lbs)
Catalog No. 50-211-2917