Learn More
CARESTREAM HEALTH INC Manual Developer & Replenisher 10 Liters 2/Pk (2x5L)
Supplier: CARESTREAM HEALTH INC 5158613
Chemical concentrate designed for manual (hand tank) processing of dental X-ray film.
This item is not returnable.
View return policy