Learn More
Med Vet International Non-Woven Gauze Sponges, 3 in. x 3 in., 4-Ply, 200/pkg
Supplier: Med Vet International PK334
Unique rayon/polyester formed fabric Significantly less linting than cotton gauze Virtually non-adherent More cost-effective than cotton gauze Non-sterile