Learn More
Nasco Gaumard™ Breat Phantom Simulator
Gaumard™ Breast Phantom Simulator provides practice for performing ultrasound identification of cysts.
Supplier: Nasco SB48847L
Description
- Left and right breasts attach to adult upper torso
- Left breast permits ultrasound identification of cysts versus dense masses, while the right breast permits identification of cysts of different sizes and depths
- Breasts easily removed and assembled
- Cysts used to teach ultrasound-guided needle aspiration
- Dense masses used for ultrasound-guided needle biopsies
- Left breast contains six dense masses and three cysts randomly oriented
- Realistic texture and look
- Right breast contains 10 cysts of different sizes and depths
- Self-healing skin
- Use in either the upright or reclining position
- Visualize cysts and masses using REAL ultrasound equipment
- Includes instruction manual and soft carry bag
Specifications
Medical Model | |
Ultrasound Training | |
Light Tan | |
Instruction manual and soft carry bag |