Learn More
Cole-Parmer Wika 213.53 2.5 0 to 3000 psi Industrial Gauge with Bottom Connection, 2 1/2" Dial
Supplier: Cole-Parmer 6804622
Gauges feature a copper alloy tube, brass socket, 304 stainless steel case, and plastic lens. All connections are 1/4" NPT(M). Shpg wt 1 lb (0.5 kg).