Learn More
Global Resources Inc GASKET (BLENDER)
Supplier:  Global Resources Inc 009762
Encompass_Preferred
Waring part number 009762 is a neoprene base gasket for Waring stainless-steel containers CAC33, SS610, SS115.  It seals the base of the container to the container adapter.  Waring considers the dimensional information as proprietary so we cannot supply that information.
Catalog No. NC1777561
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.