Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area Multitube Frame, 150-mm Scale; Two Tube, Aluminum
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX0321850
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs. Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require. These multitube frames with valves allow you to increase or decrease gas concentrations accurately at any time during an experiment¿blend two or three gases at the exact concentration you require. Frames include an installation tool and mixing tube. Note: use only with the 150-mm correlated flowtubes. Flow tubes come with gas correlation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.