Learn More
Ace Glass, Inc. 5mm OD x 210mm porosity C filter stick
Supplier: Ace Glass, Inc. 943538
5mm O.D. x 210mm gas dispersion tube for fine dispersion. Porous tip is approximately 5mm O.D. x 10mm. Supplied with porosity C (25-50 micron) filter stick.