Learn More
Uline Gas Cylinder Rack
Supplier: Uline H9680
6 cylinder capacity steel rack with individual compartments for upright storage, 11-gauge 2"""" square steel tubing, powder-coat finish, chains secure cylinders up to 12"""" diameter