Learn More
Chemglass Life Sciences Gas Collection Tube, Minum-Ware
Supplier: Chemglass Life Sciences MW4701
Gas Collection Tube, Minum-Ware. Tube consists of 15in length of 1/16in O.D. x 1/32in I.D. PTFE tubing with a 19 gauge needle inserted into both ends. The needles have a 22 bevel for septum penetration. Needle permits insertion directy into the MW-80 3 or 5ml conical reaction vial while the opposite end is inserted into a gas collection reservoir.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.