Learn More
Cole-Parmer Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 0.75 cfm, 115 VAC
Supplier: Cole-Parmer 7920030
Gardner Denver Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 0.75 cfm, 115 VAC, Wetted parts are coated with PTFE to handle your toughest applicationsWetted parts are coated with PTFE to handle your toughest applications.Lightweight aluminum construction and compact size make pumps ideal for benchtop applications such as vacuum filtration.Pumps with gauges and regulators give you more control of your vacuum/pressure applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.