Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High Purity Standards Gallium at 10,000 µg/mL in 4% HNO3 | 100mL

Supplier:  High Purity Standards 10M191100

Encompass_Preferred

Gallium (Ga) ICP Single-Component Standard at 10,000 µg/mL in 4% HNO3.

Catalog No. NC1537855


Add to Cart

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit