Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Galena Mineral Specimen
SCI_ED

For use as tool to explain specific gravity to students - EISCO Galena Mineral Specimen is used in classroom identification labs and exercises, correlates well to Next Generation Science Standards Evidence Statements.

Manufacturer:  Eisco™ ESNG0015

 View more versions of this product

Catalog No. S24687


Add to cart

Description

Description

  • Far denser than most minerals
  • Reflective cubic crystals offer shining example of metallic luster
  • Comes in clear labeled and barcoded bag for storage
Specifications

Specifications

Each
Study of specific gravity
Approx. 1 in.
Clear labeled and barcoded bag
Galena Mineral Study Specimen
Approx. 3 cm
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit