Learn More
AnaSpec GAD65 (206 - 220) TYEIAPVFVLLE
Supplier: AnaSpec AS61515
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
TYEIAPVFVLLEYVT - A Glutamic acid decarboxylase 2 (GAD2) peptide corresponding to residues 206 to 220 of GAD65.