Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec GA100 Stainless Steel Autopsy Cart Top
SDP

Mopec™ GA100 Stainless Steel Autopsy Cart Top is ideal for autopsy use and suitable for use with various Mopec™ manufactured carts.

Supplier:  Mopec GA100

Catalog No. 22-444-086


Add to cart

Description

Description

  • Type 304 stainless steel 11 gauge
  • Nylon roller wheel assembly
  • Modular construction
  • 2 Body tray bumpers per bay
  • Includes one 32 inch body tray per bay
  • Heavy duty adjustable leveling pads

Warranty and Services

1 Year Warranty

Specifications

Specifications

500 lb. per bay
80.5 in.
204.47 cm
One 32 in. Body Tray per bay
Roller Type
Stainless Steel
32 in.
81.28 cm
226.8kg per bay
Standard with no trough
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit