Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coast G32 Pure Beam Focusing Flashlight
The G32 LED Flashlight is slender with a pocket-friendly size, while still harnessing Coast's Pure Beam Focusing Optic System and the ability to twist focus.
Supplier: Coast 20484
Description
- Equipped with Coast's Pure Beam Focusing Optic System and the ability to twist focus
- While in flood mode, the flashlight casts a considerable circle of light that is equally bright all around without any dark spots
- In spot mode, the light will reach far downrange with a transition halo that allows for a large viewing area surrounding the tight spot beam
- Impact resistant and weather resistant with a virtually unbreakable LED
- A useful pocket clip is included for easy storage and transportation
- Backed by Coast’s lifetime warranty against defects in materials and workmanship
- Tested and rated to ANSI/FL1 Standards
Specifications
Flashlight | |
1.5 V | |
159 g | |
239 ft. (Low), 433 ft. (High) | |
Black | |
2 x AA Batteries, Pocket Clip | |
Lifetime |
LED | |
5.6 oz. | |
2 x AA (Alkaline) | |
ANSI FL1 | |
Pure Beam Focusing | |
110 lm (Low), 355 lm (High) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction