Learn More
Epoch Life Science Inc EconoSpin® prepacked G25 Spin Columns with inert support for Dye Terminator Removal, Desalting, Pre-filtration etc. With screw cap and snap off end-tail (50 columns)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Epoch Life Science Inc 2090050
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
PP housing with prepacked G25 on an inert support. 700 ul bed volume. 50-200 ul sample volume. Liquid tight screw cap enables long term buffer storage and eliminates splash when opening/closing. Ideal for protein desalting or post-sequencingg cleanup for Sanger sequencingg. Ideal for small volume batch mode His-tag protein purification when used with Ni+ agarose. 50 columns.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.