Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G10, >98%, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Activator of STING-dependent signaling; exhibits no direct binding affinity for STING. Stimulates IRF-3-induced transcription and induces IFNα and IFNγ gene expression in human fibroblasts. Exhibits no effect on NF-κB signaling. Blocks viral replication in vitro.
Specifications
Linear Formula | C21H16ClFN2O3S |
Quantity | 50 mg |
Solubility Information | Soluble to 100mM in DMSO |
Formula Weight | 430.88 |
Chemical Name or Material | 4-[(2-Chloro-6-fluorophenyl)methyl]-N-(2-furanylmethyl)-3,4-dihydro-3-oxo-2H-1,4-benzothiazine-6-car |
Grade | >98% |
Safety and Handling
Recommended Storage : Store at −20°C