Learn More
Perkin Elmer US LLC Furnace Shield - Platinum
Supplier: Perkin Elmer US LLC 03191628
This shield slides over the TGA 8000 furnace assembly protecting the analyzer and surrounding parts from excessively high temperatures. With this part number, you can order a replacement or spare shield.