Learn More
Chemglass Life Sciences Funnel, Vial, PTFE, 1.0" OD, 9-425 and 10-425 GPI Thread
Supplier: Chemglass Life Sciences CG176102
Funnel, Vial, PTFE, 1.0" OD, 9-425 and 10-425 GPI Thread. Funnel machined from PTFE screws securely onto any standard laboratory vial having a GPI thread. Design allows for easy, safe and mess free transfer of product.