Learn More
Chemglass Life Sciences Funnel, Vial, PTFE, 1.4" OD, 15-425 GPI Thread
Supplier: Chemglass Life Sciences CG176104
Funnel, Vial, PTFE, 1.4" OD, 15-425 GPI Thread. Funnel machined from PTFE screws securely onto any standard laboratory vial having a GPI thread. Design allows for easy, safe and mess free transfer of product.PLEASE NOTE: VIALS ARE NOT INCLUDED WITH FUNNELS.