Learn More
Chemglass Life Sciences 250mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Stopcock, 375mm OAH
Supplier: Chemglass Life Sciences CG171203
250mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Stopcock, 375mm OAH. Cylindrical style addition funnel similar to CG-1710, but with a PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.