Learn More
Ace Glass, Inc. Funnel, addition, graduated, metering stopcock, 50mL, 14/20
SDP
Supplier:  Ace Glass, Inc. 949310
Encompass_Preferred
50mL pressure equalizing addition funnel with PTFE metering stopcock. Features include 14/20 standard taper joints, protected drip-tip, equalizing arm, and double scale graduations calibrated from the stopcock.
Catalog No. 50-976-950