Learn More
Bioworld Fungigone™ Fungal Contamination Inhibitor, 100 mL
Supplier: Bioworld 406000061
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
FungiGone™FungiGone™ is bioWORLD's proprietary antifungal reagent formulationThis product drastically reducees the growth of fungi in your tissue culture applications, without affecting the plant tissue. Excellent for tissue explants for vegetative propagation, and seed germination testing.Supplied as 100X concentrateIn vitro plant tissue culture-tested
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.