Learn More
Cole-Parmer TEL UK Air Flow Monitor for Fume Hoods, 0 to 1000 fpm Airflow Monitor
Supplier: Cole-Parmer 3355660
With a reliable, durable sensor element, monitor provides simple field calibration. Multiple alarmsaudible, visual, and relay.