Learn More
Medchemexpress LLC HY-W020768 10mg Medchemexpress, Fullerene-C60 CAS:99685-96-8 Purity:>98%
Supplier: Medchemexpress LLC HYW02076810MG
Medchemexpress, HY-W020768 10mg Fullerene-C60 CAS:99685-96-8 Fullerene-C60, a representative of carbon nanocompounds, is suggested to be promising agent for application in photodynamic therapy due to its unique physicochemical properties. Fullerene-C60 probes the intramolecular dynamics of its electron and energy transfer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.