Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Impacto™ Full Finger Anti-Fatigue Thermo Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Description
- Natural pain relief for prevention of injuries during work or play
- Designed to allow full ventilation during repetitive tasks, for extended periods of time
- Light but firm therapeutic compression counteracts tissue swelling to provide temporary relief from repetitive strain injuries (RSI)
- Promotes optimal muscle function at 1°C higher than the body’s normal resting temperature
- Ideal for repetitive jobs such as sewing, or for wearing inside other gloves when handling frozen products
Specifications
Color | Black |
Material | Nylon |
Size | 2X-Large |
Finger Style | Full |
For Use With (Application) | Temporary Pain Relief from Repetative Strain Injuries (RSI) |
Glove Cuff Style | Hook and Loop |
Series | TS299 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction