Learn More
Perkin Elmer US LLC FT-IR Nutraceutical Compliance Resource Pack
Supplier: Perkin Elmer US LLC L1608005
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The FT-IR Nutraceutical Compliance Resource Pack facilitates FT-IR spectroscopic analysis for Nutraceutical and Traditional Medicine professionals. The pack covers the requirements of seven key Pharmacopoeias (USP, BP, EP, IP, JP, CP and IntPhr). The quality assessment of most of natural products is supported by an included nutraceutical starter set of spectra. The Nutraceutical Compliance Resource Pack: Facilitates rapid testing and release of samples in accordance with specific Pharmacopeia regulations, increasing lab productivity in FT-IR sampling methods and Pharmacopeia best practice. Provides quality assurance for nutraceutical ingredients. Provides training and skills development through tailored experiments.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.