Learn More
Electron Microscopy Sciences Formalin Prefilled HistoTainer, 45 ml
Supplier:  Electron Microscopy Sciences 1574390
Encompass_Preferred
The HistoTainer is half filled with 10% Neutral Buffered Formalin as a fixative. These ready-to-use prefilled containers are timer saving and make your work more efficient.
Catalog No. 50-190-1171