Learn More
Chemglass Life Sciences Fritted Disc, 10mm, Medium porosity, Filter
Supplier: Chemglass Life Sciences CG20102
Fritted Disc, 10mm, Medium porosity, Filter. Porous discs for use in fabrication of special glass apparatus.Pressure Limits:Fritted glassware is designed for low pressure vacuum filtration or gas flow. The MAX differential pressure should not exceed 15 PSI.Thermal Limits:Fritted glassware should not be exposed to excessive temperature change or direct exposure to flame due to the fact that it has less resistance to thermal shock than standard borosilicate glassware.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.