Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Frits for Isolation Technologies Precolumn Filter

Minimizes dead volume and band broadening, to prevent loss of column efficiency while protecting the column

Supplier:  MilliporeSigma™ Supelco™ 57677

 View more versions of this product

Catalog No. 11-102-0926


Add to Cart

Description

Description

  • In-line installation
  • High capacity inlet filter, with 0.5 μm stainless steel frit
  • Minimizes dead volume and band broadening
  • Prevents loss of column efficiency while protecting the column
  • Includes: Tubing (2), Nuts (2), and Ferrules (2)
Specifications

Specifications

3 mm
Stainless Steel
0.5 μm
Isolation Technology Precolumn Filter
10/Pk.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit