Learn More
Thermo Scientific FRITS 10UM
Supplier: Thermo Scientific 049453
Use these Thermo Scientific™ Dionex™ ASE™ Extraction Cell Frits to prevent plugging of sample particulates in the extraction cell.Accelerated Solvent Extractors feature extraction cells with finger-tight cell caps with compression seals for high-pressure closure and maximum ease of use. 10 um SST, ASE 200 Cells, Pkg of 50