Learn More
MilliporeSigma™ Supelco™ Frit for Omnifit™ Column Endpiece
Frit's pore diameter and surface structures should be optimized to avoid clogging and abrasion of chromatographic particles.
Supplier: MilliporeSigma™ Supelco™ 56269U
Description
- End pieces are equipped with porous frits
- Frits can be made of sintered metal, glass and plastic
- Designed to be easily exchangeable
- Can also be used to keep packing material in place
Specifications
Columns - 6.6 mm Bore | |
20 Pk. | |
25 μm |
Polyethylene | |
Frit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Omnifit is a registered trademark of Diba Industries