Learn More
MilliporeSigma™ Supelco™ Frit for Omnifit™ Column Endpiece
Optimized to avoid clogging and abrasion of chromatographic particles
Supplier: MilliporeSigma™ Supelco™ 56129
Description
- End pieces are equipped with porous frits
- Frits can be made up of sintered metal, glass or plastic
- Can also be used to keep packing material in place
- Easily exchangeable
Specifications
25 mm Bore | |
20 Pk. | |
25 μm |
PTFE | |
Frit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Omnifit is a registered trademark of Diba Industries