Learn More
Fine Science Tools Friedman-Pearson Rongeurs, Stainless Steel, 14 cm
Supplier: Fine Science Tools 1602014
Tip Shape: Straight/ Cup Size: 1mm/ Alloy / Material: Stainless Steel/ Length: 14cm/ Joint Action: Single