Learn More
Caron Products And Services Freeze/Thaw Chamber, 25 cu. ft., Temp
Supplier: Caron Products And Services 7901252
25 ft3 (708 L) Freeze/Thaw Chamber offers programmable temperature (-25C to 70C) settings. Cycles between setpoints, enabling continuous simulation of freezing and thawing for rapid testing. Includes earth-friendly and customer convenient features. 208/230V, 60 Hz.