Learn More
Sklar Instruments Freeway Graves Vaginal Speculum, Sklar, 4 in. x 1-1/4 in., Angled (90 Degrees), Open Side (Right), Smooth. EA/1.
Supplier: Sklar Instruments 903750
Freeway Graves Vaginal Speculum, Sklar, 4 in. x 1-1/4 in., Angled (90 Degrees), Open Side (Right), Smooth. The Sklar Freeway-Graves Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix. The Freeway-Graves Vaginal Speculum features an open side which allows improved visibility and more room for supplemental instrument access. It is available with 45 or 90 degree angled blades. This product is a medium sized speculum with 4 in. X 1-1/4 in. blades angled 90 degrees.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.