Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area Single-Tube Frame, 65-mm Scale; No Valve Port, Aluminum
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0322000
Encompass_Preferred
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs. Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require. Choose a frame with a valve port if you plan on adding a valve cartridge to control flow.
Catalog No. 50-627-240