Learn More
GENOVIS INC FRAGIT Z KIT 5X0.5MG
Supplier: GENOVIS INC A2FZ2025
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
FragIT Z kit, Digest and Purify up to 5 x 0.5 mg FragIT Z kit, Digest and Purify up to 5 x 0.5 mgFragIT Z kit for creation and isolation of F(ab’)2 and Fc fragments from 5 x 0.5 mg mouse IgG2a. The kit consist of two spin columns – one for fragmentation (FragIT Z column) and one for purification of F(ab’)2 and isolation of Fc fragments.The FragIT Z column is FabRICATOR Z enzyme immobilized on agarose and the second column has affinity ligands selective for Fc fragments.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.