Learn More
GENOVIS INC FRAGIT KIT CLEAVE/PURIFY 10MG
Supplier: GENOVIS INC A2FR2100
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
FragIT™ Kit MicroSpin. 10 ml1 settled resin of FabRICATOR® (IdeS) immobilized on highly cross-linked agarose. Storage temp 4-8°C. White resin. Sterility - Absence of bacterial growth3. Stability - 1 year at 4-8°C
This item is not returnable.
View return policy