Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Safety Apparel™ FR Food Processing Lab Coat
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- CAT 2
- Fabric/Material: 7 oz. UltraSoft™
- Created for food service: no buttons, no loose or hanging components
- Side pass-through pockets
- Generous cut allows for unrestricted movement
- Covered snaps on front and cuffs
- Dual certified for arc flash and flash fire protection
- Lightweight and breathable design
- FR fabric will not melt, drip, or ignite
- Made in the USA
Specifications
Size | Small |
Color | Navy |
Closure Type | Snaps |
Arc Flash Rating | 8 cal/cm2 |
Certifications/Compliance | NFPA 70E for Arc Flash, UL Certified to NFPA 2112 for Flash Fire, ASTM F1506 for Arc Flash, CSA Z462 for Arc Flash (Canada), OSHA 1910.269 |
For Use With (Application) | Food Processing |
Length (English) | 42 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction