Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Formation of Tin Wool Chemistry Demo Kit

Single replacement reaction in which zinc metal dissolves in solution and tin metal is formed.
Supplier: Innovating Science™ IS7032
Alert:
DOT Info: UN1760, Corrosive liquid, n.o.s., (Stannous Chloride, Hydrochloric Acid), 8, PG III, Ltd Qty
Includes: Tin (II) Chloride, 1.0 Solution UN1760 and 1 x 140g Zinc Metal Mossy
Description
Single replacement reaction in which zinc metal dissolves in solution and tin metal is formed.Specifications
White | |
13 to 20 | |
Tin (II) Chloride, 1.0 Solution UN1760 and 1 x 140g Zinc Metal Mossy | |
1 |
7 to 14 | |
Demonstrates how zinc metal dissolves in solution to form tin metal. | |
Formation of Tin Wool Chemistry Demo Kit | |
9.75 x 7.125 x 12.25 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title