Learn More
Electron Microscopy Sciences Formalin Prefilled HistoTainer, 60 ml
Supplier: Electron Microscopy Sciences 15743120
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The HistoTainer is half filled with 10% Neutral Buffered Formalin as a fixative. These ready-to-use prefilled containers are timer saving and make your work more efficient.