Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Formalin Jellyfish
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
For use in study of anatomy of cnidarians using common, formalin-preserved jellyfish. Carolina™ Formalin Jellyfish is shipped in Carosafe™, Carolina's proprietary shipping and holding fluid.
$4.10 - $4.45
Specifications
Type | Preserved Specimen |
---|---|
Preservative | Formalin |
Injection Type | Plain |
Packaging | Jar |
Description
- Common marine jellyfish - Aurelia
- Class Scyphozoa
- Carosafe™ is odorless fluid designed to minimize unpleasant odor of formaldehyde
Specifications
Preserved Specimen | |
Formalin | |
Plain | |
Jar |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title