Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Formalin Crayfish
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
For use in exploring relationship between structure, function, and environment using formalin-preserved crayfish. Carolina™ Formalin Crayfish is ideal invertebrate dissection specimen.
$17.25 - $24.25
Specifications
Type | Preserved Specimen |
---|---|
Preservative | Formalin |
Length (English) | 4+ in. |
Length (Metric) | 10.1+ cm |
Quantity | 10/Bag |
Description
- Pacifastacus sp.
- Phylum Arthropoda, class Crustacea
- Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
- Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde
Specifications
Preserved Specimen | |
4+ in. | |
10/Bag |
Formalin | |
10.1+ cm | |
Bulk Bag |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title