Learn More
Electron Microscopy Sciences Formaldehyde Spill Response Kit in Hard case
Supplier: Electron Microscopy Sciences 24141
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This kit features FSC-1 to aid in reducing exposure to formaldehyde gas in the event of a spill. The formalin is transformed into an easily removable semi solid mass. The resulting mass cools rapidly to minimize formaldehyde off gassing. Kits Contains 1-11oz bottle of FSC-1 solidifier, 1Pair vented goggles, 1Respirator, 1Open backed gown to protect clothing, 1Pair nitrile gloves, 4Scoop/scrapers, 3Wiper pads, and 4Thirty gallon bags.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.