Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Formaldehyde Solution ACS, MilliporeSigma™
Click to view available options
Quantity:
450mL
216kg
Packaging:
Metal Drum w/bag
Poly Bottle
Specifications
Linear Formula | HCHO |
Packaging | Poly Bottle |
Quantity | 450mL |
Formula Weight | 30.03 |
Grade | ACS |
Chemical Name or Material | Formaldehyde Solution |