Learn More
Electron Microscopy Sciences Forensic Certified SEM Mount 8 mm pin
Supplier: Electron Microscopy Sciences 7647550
This mount is made from a special certified Aluminum alloy which does not contain any of the objectionable elements that may interfere with samples collected for GSR . These mounts are 12.7 mm x 11 mm.